Learn More
Electron Microscopy Sciences Millonig's Phosphate Buffer 0.2M, 1 L
SDP
Catalog No. 50980227
Encompass_Preferred
The pH of this buffer is 7.4 (0.1M); the osmolarity is 440 mosmols, so this buffer is hypertonic to most body fluids. Ingredients: Sodium Phosphate, monobasic, Sodium Phosphate, dibasic, and 0.5% Sodium Chloride.
Catalog No. 50-980-227 Supplier Electron Microscopy Sciences Supplier No. 1158210
The pH of this buffer is 7.4 (0.1M); the osmolarity is 440 mosmols, so this buffer is hypertonic to most body fluids. Ingredients: Sodium Phosphate, monobasic, Sodium Phosphate, dibasic, and 0.5% Sodium Chloride.