Learn More
Electron Microscopy Sciences Millonig's Phosphate Buffer, 0.2M, 7.4 pH 500 ML
SDP
Catalog No. 50980226
Encompass_Preferred
The pH of this buffer is 7.4 (0.1M); the osmolarity is 440 mosmols, so this buffer is hypertonic to most body fluids. Millonig recommended to use this buffer for the fixation of very hydrated tissues with osmium tetroxide. For marine organisms, a higher concentration of sodium chloride (3%) is recommended. We supply it in a double strength (0.2M), pH 7.4. Dilution is needed before use.
pH: 7.4
Catalog No. 50-980-226 Supplier Electron Microscopy Sciences Supplier No. 1158205
The pH of this buffer is 7.4 (0.1M); the osmolarity is 440 mosmols, so this buffer is hypertonic to most body fluids. Millonig recommended to use this buffer for the fixation of very hydrated tissues with osmium tetroxide. For marine organisms, a higher concentration of sodium chloride (3%) is recommended. We supply it in a double strength (0.2M), pH 7.4. Dilution is needed before use.
pH: 7.4