Learn More
EMD MILLIPORE 3P BIOPHARM MILLPK 100 .22 GAMMA 3/4 3PK
Supplier:  EMD MILLIPORE 3P BIOPHARM MPGL1GCF3
Encompass
Stacked disc filter sterilized by gamma radiation, Inlet/Outlet: 19 mm, Filtration Area: 500 sq. cm, Water Flow Rate: 11 L/min at 2.8 bar DP, Filter Pore Size: 0.22 micrometer, Wettability: Hydrophilic, 3/PK, Filter Material: PVDF
Catalog No. NC9992428