Learn More
MilliporeSigma MILLSTAK UPOD FILTR B1HC GD3PK
Supplier:  MilliporeSigma MB1HC23CL3
Encompass
Millistak+ Depth Filter in uPod format, B1HC media series, 23 cm 2 surface area, Luer fitting connection
Catalog No. NC0269762