Learn More
Apexbio Technology LLC Milnacipran HCl 101152-94-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092314
Encompass_Preferred
Milnacipran hydrochloride (CAS 101152-94-7) is a small molecule classified as a serotonin-norepinephrine reuptake inhibitor (SNRI) It acts by selectively inhibiting the reuptake of both serotonin and norepinephrine in the central nervous system thereby modulating synaptic neurotransmitter levels Milnacipran HCl has been investigated for its ability to influence neurochemical pathways implicated in pain processing and mood regulation It is utilized in biomedical research to study the pathophysiology of fibromyalgia and related disorders involving monoaminergic dysfunction
Catalog No. 50-000-92314 Supplier Apexbio Technology LLC Supplier No. B21125.1
Milnacipran hydrochloride (CAS 101152-94-7) is a small molecule classified as a serotonin-norepinephrine reuptake inhibitor (SNRI) It acts by selectively inhibiting the reuptake of both serotonin and norepinephrine in the central nervous system thereby modulating synaptic neurotransmitter levels Milnacipran HCl has been investigated for its ability to influence neurochemical pathways implicated in pain processing and mood regulation It is utilized in biomedical research to study the pathophysiology of fibromyalgia and related disorders involving monoaminergic dysfunction