Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Milnacipran hydrochloride | 101152-94-7 | 99.7% | 282.81 | C15H23ClN2O | 10 MG
SDP

Catalog No. 502082391
Encompass_Preferred

Milnacipran hydrochloride is the hydrochloride salt of milnacipran, a serotonin-norepinephrine reuptake inhibitor supplied as a research-grade chemical for preclinical and analytical use. The compound is provided as a solid intended for laboratory applications.

  • High purity, 99.7%.
  • Molecular weight 282.81 g/mol.
  • Chemical formula C15H23ClN2O.
  • CAS number 101152-94-7.
  • Provided in a 10 mg package for laboratory use.
  • Solid form; store at 4°C, sealed and protected from moisture.

Catalog No. 50-208-2391 Supplier Medchemexpress LLC Supplier No. HYB0168A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Milnacipran hydrochloride is the hydrochloride salt of milnacipran, a serotonin-norepinephrine reuptake inhibitor supplied as a research-grade chemical for preclinical and analytical use. The compound is provided as a solid intended for laboratory applications.

  • High purity, 99.7%.
  • Molecular weight 282.81 g/mol.
  • Chemical formula C15H23ClN2O.
  • CAS number 101152-94-7.
  • Provided in a 10 mg package for laboratory use.
  • Solid form; store at 4°C, sealed and protected from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.