Learn More
Medchemexpress LLC Milnacipran hydrochloride | 101152-94-7 | 99.7% | 10 MM x 1 ML
SDP
Catalog No. 5000407194
Encompass_Preferred
Milnacipran hydrochloride is an orally active Serotonin and Norepinephrine reuptake inhibitor. It inhibits monoamine transporters, specifically the norepinephrine transporter and the serotonin transporter (Ki values of 31 and 8.5 nM, respectively). It also inhibits pERK1/2 activation.
  • Antidepressant, anxiolytic, and analgesic properties.
  • Inhibits biting behavior in mice.
  • Can be used in the study of major depressive disorder, anxiety disorders, and neuropathic pain (e.g., fibromyalgia).
Catalog No. 50-004-07194 Supplier Medchemexpress LLC Supplier No. HYB0168A10MM/1ML
Milnacipran hydrochloride is an orally active Serotonin and Norepinephrine reuptake inhibitor. It inhibits monoamine transporters, specifically the norepinephrine transporter and the serotonin transporter (Ki values of 31 and 8.5 nM, respectively). It also inhibits pERK1/2 activation.
  • Antidepressant, anxiolytic, and analgesic properties.
  • Inhibits biting behavior in mice.
  • Can be used in the study of major depressive disorder, anxiety disorders, and neuropathic pain (e.g., fibromyalgia).