Learn More
Medchemexpress LLC Milnacipran hydrochloride | 101152-94-7 | 99.7% | 100 MG
SDP
Catalog No. 5000406080
Encompass_Preferred
Milnacipran hydrochloride is an orally active serotonin and norepinephrine reuptake inhibitor. It inhibits monoamine transporters, specifically the norepinephrine transporter and the serotonin transporter, with Ki values of 31 nM and 8.5 nM, respectively. It also inhibits pERK1/2 activation. This compound possesses antidepressant, anxiolytic, and analgesic properties, and has been observed to inhibit biting behavior in mice.
  • It is an orally active serotonin and norepinephrine reuptake inhibitor.
  • It inhibits monoamine transporters, with notable activity on norepinephrine and serotonin transporters.
  • The compound inhibits pERK1/2 activation.
  • It demonstrates antidepressant, anxiolytic, and analgesic properties.
  • It can be used in studies related to major depressive disorder, anxiety disorders, and neuropathic pain, including fibromyalgia.
Catalog No. 50-004-06080 Supplier Medchemexpress LLC Supplier No. HYB0168A100MG
Milnacipran hydrochloride is an orally active serotonin and norepinephrine reuptake inhibitor. It inhibits monoamine transporters, specifically the norepinephrine transporter and the serotonin transporter, with Ki values of 31 nM and 8.5 nM, respectively. It also inhibits pERK1/2 activation. This compound possesses antidepressant, anxiolytic, and analgesic properties, and has been observed to inhibit biting behavior in mice.
  • It is an orally active serotonin and norepinephrine reuptake inhibitor.
  • It inhibits monoamine transporters, with notable activity on norepinephrine and serotonin transporters.
  • The compound inhibits pERK1/2 activation.
  • It demonstrates antidepressant, anxiolytic, and analgesic properties.
  • It can be used in studies related to major depressive disorder, anxiety disorders, and neuropathic pain, including fibromyalgia.