Learn More
Medchemexpress LLC Milnacipran hydrochloride | 101152-94-7 | 99.7% | 25 MG
SDP
Catalog No. 5000450979
Encompass_Preferred
Milnacipran hydrochloride is an orally active Serotonin and Norepinephrine reuptake inhibitor. It inhibits monoamine transporters, specifically the norepinephrine transporter and the serotonin transporter, with Ki values of 31 and 8.5 nM, respectively. It also inhibits pERK1/2 activation. Milnacipran hydrochloride exhibits antidepressant, anxiolytic, and analgesic properties and can inhibit biting behavior in mice. It is used in the study of major depressive disorder, anxiety disorders, and neuropathic pain, such as fibromyalgia.
  • Orally active serotonin and norepinephrine reuptake inhibitor
  • Inhibits monoamine transporters with Ki values of 31 and 8.5 nM
  • Inhibits pERK1/2 activation
  • Exhibits antidepressant, anxiolytic, and analgesic properties
  • Can inhibit biting behavior in mice
  • Used in studies for major depressive disorder, anxiety disorders, and neuropathic pain
Catalog No. 50-004-50979 Supplier Medchemexpress LLC Supplier No. 25MGHYB0168A
Milnacipran hydrochloride is an orally active Serotonin and Norepinephrine reuptake inhibitor. It inhibits monoamine transporters, specifically the norepinephrine transporter and the serotonin transporter, with Ki values of 31 and 8.5 nM, respectively. It also inhibits pERK1/2 activation. Milnacipran hydrochloride exhibits antidepressant, anxiolytic, and analgesic properties and can inhibit biting behavior in mice. It is used in the study of major depressive disorder, anxiety disorders, and neuropathic pain, such as fibromyalgia.
  • Orally active serotonin and norepinephrine reuptake inhibitor
  • Inhibits monoamine transporters with Ki values of 31 and 8.5 nM
  • Inhibits pERK1/2 activation
  • Exhibits antidepressant, anxiolytic, and analgesic properties
  • Can inhibit biting behavior in mice
  • Used in studies for major depressive disorder, anxiety disorders, and neuropathic pain