Learn More
Medchemexpress LLC Milnacipran hydrochloride | 101152-94-7 | 99.7% | 5 MG
SDP
Catalog No. 5000405348
Encompass_Preferred
Milnacipran hydrochloride is an orally active serotonin and norepinephrine reuptake inhibitor. It specifically inhibits monoamine transporters with Ki values of 31 nM for the norepinephrine transporter and 8.5 nM for the serotonin transporter. This compound also inhibits pERK1/2 activation and exhibits antidepressant, anxiolytic, and analgesic properties. It is used in the study of major depressive disorder, anxiety disorders, and neuropathic pain, including fibromyalgia.
  • Inhibits norepinephrine and serotonin transporters.
  • Inhibits pERK1/2 activation.
  • Possesses antidepressant, anxiolytic, and analgesic properties.
  • Used in studies of major depressive disorder, anxiety disorders, and neuropathic pain.
Catalog No. 50-004-05348 Supplier Medchemexpress LLC Supplier No. HYB0168A5MG
Milnacipran hydrochloride is an orally active serotonin and norepinephrine reuptake inhibitor. It specifically inhibits monoamine transporters with Ki values of 31 nM for the norepinephrine transporter and 8.5 nM for the serotonin transporter. This compound also inhibits pERK1/2 activation and exhibits antidepressant, anxiolytic, and analgesic properties. It is used in the study of major depressive disorder, anxiety disorders, and neuropathic pain, including fibromyalgia.
  • Inhibits norepinephrine and serotonin transporters.
  • Inhibits pERK1/2 activation.
  • Possesses antidepressant, anxiolytic, and analgesic properties.
  • Used in studies of major depressive disorder, anxiety disorders, and neuropathic pain.