Learn More
TARGETMOL CHEMICALS INC Milnacipran hydrochloride
SDP
Catalog No. 502879640
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Milnacipran hydrochloride (Savella) is a serotonin and norepinephrine reuptake inhibitor used to treat major depressive disorders. Milnacipran hydrochloride is an enantiomer of milnacipran, which is used to treat fibromyalgia. Milnacipran hydrochloride and milnacipran have been associated with a low rate of transient elevations in serum aminotransferase levels during treatment and with rare instances of clinically apparent acute liver injury with jaundice. Purity 100%
Catalog No. 50-287-9640 Supplier TARGETMOL CHEMICALS INC Supplier No. T1189100MG
Also available in 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Milnacipran hydrochloride (Savella) is a serotonin and norepinephrine reuptake inhibitor used to treat major depressive disorders. Milnacipran hydrochloride is an enantiomer of milnacipran, which is used to treat fibromyalgia. Milnacipran hydrochloride and milnacipran have been associated with a low rate of transient elevations in serum aminotransferase levels during treatment and with rare instances of clinically apparent acute liver injury with jaundice. Purity 100%