Learn More
Abcam Milnacipran hydrochloride, Serotonin and norepinephrine reuptake inhibitor, 10MG
Catalog No. 502707307
Encompass_Preferred
MW 282.81 g/mol, Purity >99%. Serotonin and norepinephrine reuptake inhibitor. IC₅₀ values are 100, 203 and >100000 nM for NA, 5-HT and DA, respectively. Centrally active following oral administration. Exhibits antidepressant and antifibromyalgia activity in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7307 Supplier Abcam Supplier No. AB12075510MG
MW 282.81 g/mol, Purity >99%. Serotonin and norepinephrine reuptake inhibitor. IC₅₀ values are 100, 203 and >100000 nM for NA, 5-HT and DA, respectively. Centrally active following oral administration. Exhibits antidepressant and antifibromyalgia activity in vivo.
The product is subject to the following: Abcam Restricted Use Statement