Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Milnacipran Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M2133500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2133500MG 500 mg
M213350MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2133500MG Supplier TCI America Supplier No. M2133500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 101152-94-7
Molecular Formula C15H23ClN2O
Molecular Weight (g/mol) 282.812
MDL Number MFCD00901293
InChI Key XNCDYJFPRPDERF-LKNSGBSQSA-N
Synonym dalcipran, ixel, rac-milnacipran hydrochloride, 1r,2s-rel-2 aminomethyl-n,n-diethyl-1-phenyl-cyclopropanecarboxamide, monohydrochloride, 2s-2-aminomethyl-n,n-diethyl-1-phenylcyclopropane-1-carboxamide hydrochloride
PubChem CID 45039934
IUPAC Name (2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide;hydrochloride
SMILES CCN(CC)C(=O)C1(CC1CN)C2=CC=CC=C2.Cl

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 500 mg
UN Number 2811
Formula Weight 282.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Milnacipran Hydrochloride
RTECSNumber GZ1014010
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.