Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Milnacipran hydrochloride, Tocris Bioscience™

Catalog No. 328610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-861-0 10 mg
32-865-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-861-0 Supplier R&D Systems Supplier No. 3286/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

5-HT and NA reuptake inhibitor (SNRI)

Orally active 5-HT and noradrenalin re-uptake inhibitor (SNRI) (IC50 values are 203 and 100 nM respectively) that displays no affinity at a range of other receptors. Causes adaptive changes to α1-adrenergic and 5-HT2A serotonergic systems when administered repeatedly. Exhibits antidepressive and antinociceptive activities in vivo.

Chemical Identifiers

CAS 101152-94-7
Molecular Formula C15H23ClN2O
Molecular Weight (g/mol) 282.812
InChI Key XNCDYJFPRPDERF-LKNSGBSQSA-N
Synonym dalcipran, ixel, rac-milnacipran hydrochloride, 1r,2s-rel-2 aminomethyl-n,n-diethyl-1-phenyl-cyclopropanecarboxamide, monohydrochloride, 2s-2-aminomethyl-n,n-diethyl-1-phenylcyclopropane-1-carboxamide hydrochloride
PubChem CID 45039934
IUPAC Name (2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide;hydrochloride
SMILES CCN(CC)C(=O)C1(CC1CN)C2=CC=CC=C2.Cl

Specifications

Quantity 10 mg
Formula Weight 282.81
Percent Purity >99%
Chemical Name or Material Milnacipran hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.