Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Milnacipran hydrochloride, Tocris Bioscience™
Description
Orally active 5-HT and noradrenalin re-uptake inhibitor (SNRI) (IC50 values are 203 and 100 nM respectively) that displays no affinity at a range of other receptors. Causes adaptive changes to α1-adrenergic and 5-HT2A serotonergic systems when administered repeatedly. Exhibits antidepressive and antinociceptive activities in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 282.81 |
| Percent Purity | >99% |
| Chemical Name or Material | Milnacipran hydrochloride |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?