Learn More
Cayman Chemical Milrinone-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2749393-50-6, 1 mg
SDP
Catalog No. 502933253
Encompass_Preferred
An internal standard for the quantification of milrinone by GC- or LC-MS
Catalog No. 50-293-3253 Supplier Cayman Chemical Supplier No. 254291 MG
An internal standard for the quantification of milrinone by GC- or LC-MS