Learn More
Enzo Life Sciences Milrinone (5mg). CAS: 78415-72-2
SDP
Supplier:  Enzo Life Sciences ALX270083M005
Encompass_Preferred
PDE3 inhibitor. Selective inhibitor of phosphodiesterase 3 (PDE3, IC50=0.3µM). Alternative name: 1,6-Dihydro-2-methyl-6-oxo-3,4'-bipyridine-5-carbonitrile. Formula: C12H9N3O. MW: 211.2. MI: 14: 6197. Purity: ≥97% (Total Nitrogen). Solubility: Soluble in DMSO (20mg/ml) or warm 100% ethanol (3mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8400