Learn More
Medchemexpress LLC Milrinone | 78415-72-2 | 99.9% | C12H9N3O | 1 ML
SDP
Catalog No. 5000353646
Encompass_Preferred
Milrinone is a PDE3 inhibitor, as well as an inotrope and vasodilator.
  • Inhibits PDE3
  • Increases PKA activity in hypoxic myocytes to normoxic levels
  • Normalizes TP receptor sensitivity
  • Reduces NE-induced vasoconstriction
  • Reduces pulmonary artery pressure and increases cAMP plasma concentrations
  • Exhibits antiplatelet activity
Catalog No. 50-003-53646 Supplier Medchemexpress LLC Supplier No. HY1425210MM/1ML
Milrinone is a PDE3 inhibitor, as well as an inotrope and vasodilator.
  • Inhibits PDE3
  • Increases PKA activity in hypoxic myocytes to normoxic levels
  • Normalizes TP receptor sensitivity
  • Reduces NE-induced vasoconstriction
  • Reduces pulmonary artery pressure and increases cAMP plasma concentrations
  • Exhibits antiplatelet activity