Learn More
Medchemexpress LLC Milrinone | 78415-72-2 | 99.9% | C12H9N3O | 200 MG
SDP
Catalog No. 5000353647
Encompass_Preferred
Milrinone (Win 47203) is a potent Phosphodiesterase 3 (PDE3) inhibitor that functions as both an inotrope and vasodilator. By inhibiting PDE3, Milrinone increases intracellular cyclic adenosine monophosphate (cAMP) levels, leading to strengthened heart muscle contractions and widened blood vessels. This product is for research use only.
  • Targets phosphodiesterase 3 (PDE3) as an inhibitor
  • Acts as an inotrope and vasodilator
  • Increases PKA activity in hypoxic myocytes
  • Reduces norepinephrine-induced vasoconstriction
  • Attenuates pulmonary hypertension in animal models
  • Exhibits antiplatelet activity
Catalog No. 50-003-53647 Supplier Medchemexpress LLC Supplier No. HY14252200MG
Milrinone (Win 47203) is a potent Phosphodiesterase 3 (PDE3) inhibitor that functions as both an inotrope and vasodilator. By inhibiting PDE3, Milrinone increases intracellular cyclic adenosine monophosphate (cAMP) levels, leading to strengthened heart muscle contractions and widened blood vessels. This product is for research use only.
  • Targets phosphodiesterase 3 (PDE3) as an inhibitor
  • Acts as an inotrope and vasodilator
  • Increases PKA activity in hypoxic myocytes
  • Reduces norepinephrine-induced vasoconstriction
  • Attenuates pulmonary hypertension in animal models
  • Exhibits antiplatelet activity