Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Milrinone | 78415-72-2 | 99.9% | C12H9N3O | 500 MG
SDP

Catalog No. 5000353677
Encompass_Preferred

Milrinone is a potent PDE3 inhibitor, also functioning as an inotrope and vasodilator. This product is intended for research use only and is not for patient administration.

  • Potent PDE3 inhibitor
  • Functions as an inotrope and vasodilator
  • Appears as a solid, white to off-white powder
  • Soluble in DMSO at 50 mg/mL
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-53677 Supplier Medchemexpress LLC Supplier No. HY14252500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Milrinone is a potent PDE3 inhibitor, also functioning as an inotrope and vasodilator. This product is intended for research use only and is not for patient administration.

  • Potent PDE3 inhibitor
  • Functions as an inotrope and vasodilator
  • Appears as a solid, white to off-white powder
  • Soluble in DMSO at 50 mg/mL
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.