Learn More
Selleck Chemical LLC Milrinone 50mg 78415-72-2 Win 47203
SDP
Supplier:  Selleck Chemical LLC S248450MG
Encompass_Preferred
Milrinone (Win 47203) is a phosphodiesterase 3 (PDE3) inhibitor, used to increase the heart's contractility. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-551-8