Learn More
Strem, An Ascensus Company CAS 863971-62-4. 500mg. Carbonylchlorohydrido[6-(di-t-butylphosphinomethyl)-2-(NN-diethylaminomethyl)pyridine]ruthenium(II) min. 98% (Milstein Catalyst Precursor). MFCD16038117
SDP
Catalog No. 502110821
Encompass
CAS 863971-62-4. 500mg. Carbonylchlorohydrido[6-(di-t-butylphosphinomethyl)-2-(NN-diethylaminomethyl)pyridine]ruthenium(II) min. 98% (Milstein Catalyst Precursor). MFCD16038117. Molecular Weight 488.01. Molecular Formula C20H36ClN2OPRu. Color/form pale yellow solid Strem 44-0081. http//www.strem.com/catalog/v/44-0081/
Catalog No. 50-211-0821 Supplier Strem, An Ascensus Company Supplier No. 440081500MG
CAS 863971-62-4. 500mg. Carbonylchlorohydrido[6-(di-t-butylphosphinomethyl)-2-(NN-diethylaminomethyl)pyridine]ruthenium(II) min. 98% (Milstein Catalyst Precursor). MFCD16038117. Molecular Weight 488.01. Molecular Formula C20H36ClN2OPRu. Color/form pale yellow solid Strem 44-0081. http//www.strem.com/catalog/v/44-0081/