Learn More
Cayman Chemical Miltefosine, Purity: ≥98%, CAS: 58066-85-6, 50 mg
SDP
Catalog No. 502896194
Encompass_Preferred
An inhibitor of CCT; inhibits liposome-induced CCT activity in MDCK cell homogenates from 10-50 µM, as well as induces translocation of CCT from the cell membrane to the cytosol in MDCK cells; inhibits PMA-induced phosphatidylcholine biosynthesis in MDCK and HeLa cells at 50 µM; inhibits phosphatidylserine-activated PKC (IC50 = 62 µM), as well as PMA-induced morphological changes and proliferation of MDCK cells; active against clinical isolate promastigotes of L. infantum (EC50s = 5-25 µM); topical application completely eradicates L. amazonensis and induces re-epithelialization of lesions in a mouse model of cutaneous leishmaniasis at 0.5%
Catalog No. 50-289-6194 Supplier Cayman Chemical Supplier No. 6328050 MG
An inhibitor of CCT; inhibits liposome-induced CCT activity in MDCK cell homogenates from 10-50 µM, as well as induces translocation of CCT from the cell membrane to the cytosol in MDCK cells; inhibits PMA-induced phosphatidylcholine biosynthesis in MDCK and HeLa cells at 50 µM; inhibits phosphatidylserine-activated PKC (IC50 = 62 µM), as well as PMA-induced morphological changes and proliferation of MDCK cells; active against clinical isolate promastigotes of L. infantum (EC50s = 5-25 µM); topical application completely eradicates L. amazonensis and induces re-epithelialization of lesions in a mouse model of cutaneous leishmaniasis at 0.5%