Learn More
Cayman Chemical Miltefosine-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502910865
Encompass_Preferred
An internal standard for the quantification of miltefosine by GC- or LC-MS
Catalog No. 50-291-0865 Supplier Cayman Chemical Supplier No. 347071 MG
An internal standard for the quantification of miltefosine by GC- or LC-MS