Learn More
Apexbio Technology LLC Miltefosine, 50mg. Cas: 58066-85-6 MFCD: MFCD00133396. PI3K/Akt inhibitor
SDP
Supplier:  Apexbio Technology LLC B137150
Encompass_Preferred
Miltefosine inhibits PI3K/Akt activity with ED50 of 17.2 uM and 8.1 uM in carcinoma cell lines A431 and HeLa, first oral drug for Visceral leishmaniasis, effective against both promastigotes and amastigotes. Phase 4. 58066-85-6. MFCD00133396
Catalog No. 50-101-0817