Learn More
Ted Pella Inc MILTEX BIOPSY PUNCH WITH PLUN
SDP
Catalog No. NC0749050
Encompass
Miltex Biopsy Punch with Plunger, ID 2.0mm, OD 2.36mm, Green/Brown
Catalog No. NC0749050 Supplier Ted Pella Inc Supplier No. 1511020
Miltex Biopsy Punch with Plunger, ID 2.0mm, OD 2.36mm, Green/Brown