Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Biopsy Punches with Plunger System

Catalog No. 12460407
Change view
Click to view available options
Diameter (Metric):
1 mm
1.5 mm
2 mm
3 mm
4 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
12-460-407 3 mm
12-460-410 4 mm
12-460-405 2 mm
12-460-402 1 mm
12-460-403 1.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 12-460-407 Supplier Integra™ Supplier No. 3332P/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Effortless ejection of lodged specimens.

Effortless ejection of lodged specimens

  • Seamless, stainless steel with razor-sharp cutting edge to eliminate torn or pulled biopsies
  • Ribbed handle provides comfort and control
  • Single-use
  • Sterile
  • Color coded with size clearly marked on each punch

Order Info

Five-year shelf life

Specifications

Blade Material Stainless Steel
Product Type Biopsy Punch
For Use With (Application) Offer the added benefit of effortless ejection of Lodged Specimens
Handle Type Ribbed
Description Seamless
Disposable Single-use
Sterility Sterile
Includes Plunger System
Quantity 25/Pk.
Diameter (English) 0.12 in.
Diameter (Metric) 3 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.