Learn More
Medchemexpress LLC Milveterol (hydrochloride) | 804518-03-4 | 98.2% | C25H30ClN3O4 | 100 MG
SDP
Catalog No. 5000356107
Encompass_Preferred
Milveterol hydrochloride is a long-acting β2-adrenergic receptor agonist. It can be used in the study of asthma and chronic obstructive pulmonary disease (COPD).
  • Long-acting β2-adrenergic receptor agonist
  • Suitable for asthma research
  • Suitable for chronic obstructive pulmonary disease (COPD) research
  • Bronchodilator
Catalog No. 50-003-56107 Supplier Medchemexpress LLC Supplier No. HY14298100MG
Milveterol hydrochloride is a long-acting β2-adrenergic receptor agonist. It can be used in the study of asthma and chronic obstructive pulmonary disease (COPD).
  • Long-acting β2-adrenergic receptor agonist
  • Suitable for asthma research
  • Suitable for chronic obstructive pulmonary disease (COPD) research
  • Bronchodilator