Learn More
Medchemexpress LLC Milvexian | 1802425-99-5 | 99.9% | 626.44 g/mol | C28H23Cl2F2N9O2 | 5MG
SDP
Catalog No. 5000239082
Encompass_Preferred
Milvexian is an orally bioavailable, reversible small-molecule inhibitor of coagulation factor XIa intended for research use in antithrombotic and coagulation studies. The compound is supplied as a solid (white to off-white) and is accompanied by analytical documentation to support its use in preclinical and mechanistic experiments.
  • Orally bioavailable, reversible factor XIa inhibitor.
  • Molecular formula C28H23Cl2F2N9O2 and molecular weight 626.44 g/mol.
  • Reported purity 99.93%.
  • Provided as a solid, white to off-white powder.
  • Supplied in small research quantities, for example 5 mg packages.
  • Documentation available: datasheet, certificate of analysis, HNMR, LC-MS, and safety data sheet.
Catalog No. 50-002-39082 Supplier Medchemexpress LLC Supplier No. HY1258565MG
Milvexian is an orally bioavailable, reversible small-molecule inhibitor of coagulation factor XIa intended for research use in antithrombotic and coagulation studies. The compound is supplied as a solid (white to off-white) and is accompanied by analytical documentation to support its use in preclinical and mechanistic experiments.
  • Orally bioavailable, reversible factor XIa inhibitor.
  • Molecular formula C28H23Cl2F2N9O2 and molecular weight 626.44 g/mol.
  • Reported purity 99.93%.
  • Provided as a solid, white to off-white powder.
  • Supplied in small research quantities, for example 5 mg packages.
  • Documentation available: datasheet, certificate of analysis, HNMR, LC-MS, and safety data sheet.