Learn More
MSC Milwaukee BOLT Eye Visor, Clear Dual Coat, Z87.1, 1 ea
Catalog No. 5000795011
Encompass
The BOLT Eye Visor provides optimal fit and easy headlamp use. Meeting Z87.1+ safety standards, it functions as spectacles, offering an alternative to safety glasses. Its pivoting, adjustable design ensures a comfortable fit. It includes a BOLT Low-Profile Headlamp Mount. Each visor features a fog-free interior and an anti-scratch exterior coating for durability.
  • Best fit and easy headlamp use
  • More comfortable pivoting visor angle
  • Raise and lower with headlamp mounted
  • Lasting fog-free interior coating
  • Secure accessories simultaneously
  • Secures into front mount
  • Compatible with safety helmets and hard hats
  • ANSI/ISEA Z87.1 (+) - 2020 and CSA Z94.3 rated
  • Exterior anti-scratch hard coating
  • High-impact polycarbonate lens
  • Secure locking captive slide
Catalog No. 50-007-95011 Supplier MSC Supplier No. 41022880
The BOLT Eye Visor provides optimal fit and easy headlamp use. Meeting Z87.1+ safety standards, it functions as spectacles, offering an alternative to safety glasses. Its pivoting, adjustable design ensures a comfortable fit. It includes a BOLT Low-Profile Headlamp Mount. Each visor features a fog-free interior and an anti-scratch exterior coating for durability.
  • Best fit and easy headlamp use
  • More comfortable pivoting visor angle
  • Raise and lower with headlamp mounted
  • Lasting fog-free interior coating
  • Secure accessories simultaneously
  • Secures into front mount
  • Compatible with safety helmets and hard hats
  • ANSI/ISEA Z87.1 (+) - 2020 and CSA Z94.3 rated
  • Exterior anti-scratch hard coating
  • High-impact polycarbonate lens
  • Secure locking captive slide