Learn More
MSC Milwaukee BOLT Full Brim Hard Hat, Class C, 1 ea
Catalog No. 5000781660
Encompass
The Milwaukee BOLT Full Brim Hard Hat is designed to provide comprehensive head protection in demanding work environments. Featuring a full brim for enhanced coverage, this hard hat ensures wearer safety and comfort throughout the workday.
  • Class C rating for general protection against impacts.
  • Equipped with a versatile 4-point and 6-point suspension system for a secure and comfortable fit.
  • Durable construction for reliable, long-lasting performance.
  • Provides enhanced coverage against sun and falling debris.
Catalog No. 50-007-81660 Supplier MSC Supplier No. 48319677
The Milwaukee BOLT Full Brim Hard Hat is designed to provide comprehensive head protection in demanding work environments. Featuring a full brim for enhanced coverage, this hard hat ensures wearer safety and comfort throughout the workday.
  • Class C rating for general protection against impacts.
  • Equipped with a versatile 4-point and 6-point suspension system for a secure and comfortable fit.
  • Durable construction for reliable, long-lasting performance.
  • Provides enhanced coverage against sun and falling debris.