Learn More
MSC Milwaukee Cut Level 1 Nitrile Dipped Gloves, XXL, 1 Pair
Catalog No. 5000796435
Encompass
These ANSI/ISEA 105-2016 Cut Level 1 Nitrile Dipped Gloves are designed for durability and protection. They feature a soft, moisture-wicking liner and a sandy nitrile coating for improved grip and dexterity, ideal for handling small objects.
  • Soft and comfortable liner
  • Sandy nitrile coating for better grip
  • Improved dexterity for easier mobility
  • Nylon/Lycra blend for moisture wicking
  • SMARTSWIPE™ touchscreen compatible fingertips
  • Breathable glove design
  • Color-coded band for easy cut level identification
Catalog No. 50-007-96435 Supplier MSC Supplier No. 10883007
These ANSI/ISEA 105-2016 Cut Level 1 Nitrile Dipped Gloves are designed for durability and protection. They feature a soft, moisture-wicking liner and a sandy nitrile coating for improved grip and dexterity, ideal for handling small objects.
  • Soft and comfortable liner
  • Sandy nitrile coating for better grip
  • Improved dexterity for easier mobility
  • Nylon/Lycra blend for moisture wicking
  • SMARTSWIPE™ touchscreen compatible fingertips
  • Breathable glove design
  • Color-coded band for easy cut level identification