Learn More
MSC Milwaukee Cut-Resistant Gloves, Cut A1, 15 ga, 2X-Large, 1 Pair
Catalog No. 5000677199
Encompass
These cut-resistant gloves are designed for protection with a Cut A1, Puncture 0, and Abrasion 0 rating, meeting EN388 3121A standards. They are constructed with a 15 gauge knit from a polyester and HPPE blend, featuring seamless lining for comfort. The 2X-Large gloves have a polyethylene-coated palm and fingers for a rough grip, and are touchscreen compatible.
  • Cut A1, Puncture 0, Abrasion 0 ratings
  • Features 15 gauge knit for better dexterity
  • Provides a light and breathable design
  • Includes polyethylene-coated palm and fingers for rough grip
  • Equipped with Smartswipe™ touchscreen compatible fingertips
  • Designed with seamless polyester and HPPE blend lining
  • Measures 9-3/4 in. overall length
  • Comes with an elastic knit cuff
Catalog No. 50-006-77199 Supplier MSC Supplier No. 10883817
These cut-resistant gloves are designed for protection with a Cut A1, Puncture 0, and Abrasion 0 rating, meeting EN388 3121A standards. They are constructed with a 15 gauge knit from a polyester and HPPE blend, featuring seamless lining for comfort. The 2X-Large gloves have a polyethylene-coated palm and fingers for a rough grip, and are touchscreen compatible.
  • Cut A1, Puncture 0, Abrasion 0 ratings
  • Features 15 gauge knit for better dexterity
  • Provides a light and breathable design
  • Includes polyethylene-coated palm and fingers for rough grip
  • Equipped with Smartswipe™ touchscreen compatible fingertips
  • Designed with seamless polyester and HPPE blend lining
  • Measures 9-3/4 in. overall length
  • Comes with an elastic knit cuff