Learn More
MSC Milwaukee Cut-Resistant Gloves, Cut A5, 18 Gauge, Small, 1 Pair
Catalog No. 5000674766
Encompass
These cut-resistant gloves from Milwaukee Tool feature polyethylene and nitrile coating on the palm and fingers, providing a nitrile-dipped grip. Designed with a knit style and elastic cuff for comfort and protection, these 18 gauge gloves offer ANSI/ISEA cut resistance level A5, puncture level 0, and abrasion level 0, with an overall length of 10 in.
  • Knit style and elastic cuff for comfort
  • Nitrile coating on palm and fingers for grip
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 0
  • ANSI/ISEA puncture resistance level 0
  • Features 18 gauge construction
  • Rated EN 388 4X43E
  • Made from nylon and spandex
  • Overall length 10 in.
  • Unisex design
Catalog No. 50-006-74766 Supplier MSC Supplier No. 51348746
These cut-resistant gloves from Milwaukee Tool feature polyethylene and nitrile coating on the palm and fingers, providing a nitrile-dipped grip. Designed with a knit style and elastic cuff for comfort and protection, these 18 gauge gloves offer ANSI/ISEA cut resistance level A5, puncture level 0, and abrasion level 0, with an overall length of 10 in.
  • Knit style and elastic cuff for comfort
  • Nitrile coating on palm and fingers for grip
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 0
  • ANSI/ISEA puncture resistance level 0
  • Features 18 gauge construction
  • Rated EN 388 4X43E
  • Made from nylon and spandex
  • Overall length 10 in.
  • Unisex design