Learn More
MSC Milwaukee Cut-Resistant Gloves, SMARTSWIPE, Cut A3, Small, 1 Pair
Catalog No. 5000803724
Encompass
These cut-resistant gloves feature SMARTSWIPE™ technology for enhanced protection and functionality. They are designed for industrial applications requiring reliable cut, puncture, and abrasion resistance.
  • Cut A3 ANSI/ISEA resistance level
  • Puncture 0 ANSI/ISEA resistance level
  • Abrasion 0 ANSI/ISEA resistance level
  • Nitrile-coated palm and fingers for improved grip
  • SMARTSWIPE™ technology allows touchscreen use
  • Knit style with elastic cuff for comfort and secure fit
  • Fifteen gauge for optimal dexterity and protection
  • Made from durable nitrile and synthetic leather materials
  • Unisex design, size Small
  • EN 388 rating of 4343C
Catalog No. 50-008-03724 Supplier MSC Supplier No. 77168664
These cut-resistant gloves feature SMARTSWIPE™ technology for enhanced protection and functionality. They are designed for industrial applications requiring reliable cut, puncture, and abrasion resistance.
  • Cut A3 ANSI/ISEA resistance level
  • Puncture 0 ANSI/ISEA resistance level
  • Abrasion 0 ANSI/ISEA resistance level
  • Nitrile-coated palm and fingers for improved grip
  • SMARTSWIPE™ technology allows touchscreen use
  • Knit style with elastic cuff for comfort and secure fit
  • Fifteen gauge for optimal dexterity and protection
  • Made from durable nitrile and synthetic leather materials
  • Unisex design, size Small
  • EN 388 rating of 4343C