The Milwaukee BOLT Full Brim Vented Hard Hat with 6-point suspension is designed for jobsite adaptability, offering protection against top impacts (Type 1) while being conductive and vented (Class C). It integrates personal protection equipment and accessories through its headlamp mount and multiple accessory slots, featuring an adjustable ratcheting suspension and a moisture-wicking sweatband for comfort.
- Adjustable swinging ratchet for comfort
- Ratcheting suspension for quick adjustment
- Six accessory slots (four proprietary, two universal) for attachments
- Moisture-wicking sweatband and anti-microbial liner
- Cushioned ratchet and crown pads
- Head size range of 6.5–8.5 in
- HDPE shell material
- ANSI/ISEA Z89.1 Type 1 Class C certified
- CSA Z94.1 Type 1 Class C certified
- UL third-party tested and certified