The BOLT Full Face Shield provides long-lasting visibility and robust protection. Featuring a gray dual-coat polycarbonate lens, it includes a fog-free interior and anti-scratch exterior for enhanced durability. Designed for quick, tool-free attachment, this shield is compatible with safety helmets that have no brim and is part of the BOLT accessory system.
- Long-lasting visibility
- Lasting fog-free interior coating
- Anti-scratch exterior hard coating
- Quick attachment and removal
- Secures into BOLT front slots
- Compatible with safety helmets with no brim
- ANSI/ISEA Z87.1 2020 rated
- High-impact polycarbonate shield
- Compatible with safety glasses
- Wide 180° field of view
- Secure locking detents
- Provides limited eye protection from light to moderate impact hazards
- Blocks over 99.9% UVA, UVB, and UVC