Learn More
MSC Milwaukee Hard Hat, Class C, ANSI/ISEA Z89.1, 1 ea
Catalog No. 5000769186
Encompass
The Milwaukee BOLT™ Full Brim Hard Hat is designed for head protection in various work environments. This Class C hard hat offers comprehensive safety, meeting established industry standards for impact and penetration.
  • Available with 4pt and 6pt suspension for customized fit.
  • Meets ANSI/ISEA Z89.1 Type I standard.
  • Meets CSA Z94.1 standard.
  • Full brim design provides additional protection from sun and debris.
Catalog No. 50-007-69186 Supplier MSC Supplier No. 48320568
The Milwaukee BOLT™ Full Brim Hard Hat is designed for head protection in various work environments. This Class C hard hat offers comprehensive safety, meeting established industry standards for impact and penetration.
  • Available with 4pt and 6pt suspension for customized fit.
  • Meets ANSI/ISEA Z89.1 Type I standard.
  • Meets CSA Z94.1 standard.
  • Full brim design provides additional protection from sun and debris.