This full brim vented hard hat provides robust head protection, adapting to jobsite conditions with its integrated features. It offers multiple accessory slots for PPE integration, an adjustable ratcheting suspension for comfort, and a moisture-wicking sweatband for wearer comfort. This hard hat is ANSI/CSA approved.
- BOLT™ headlamp mount (compatible with most headlamps)
- Four BOLT™ and two universal accessory slots
- Adjustable swinging ratchet with 6-point suspension for comfort and quick adjustment
- Moisture-wicking, anti-microbial sweatband keeps wearer cool and dry
- BOLT™ marker clip for easy pen and marker access
- Cushioned ratchet and crown pads; largest ratcheting adjustment range (6.5 in–8.5 in head size)
- Replaceable and machine washable liner and sweatband
- Made in USA with global materials