Learn More
MSC Milwaukee Hard Hat, Green, Vented, 1 ea
Catalog No. 5000687103
Encompass
This hard hat features a 6-point ratcheting suspension and is designed to adapt to jobsite needs. It includes a BOLT™ Headlamp Mount, accessory slots, and a moisture-wicking sweatband for comfort.
  • Adjustable swinging ratchet for better comfort
  • Ratcheting suspension for quick adjustment
  • Six accessory slots for quick access
  • Moisture wicking sweatband to stay cool and dry fast
  • Secure accessories simultaneously
  • Cushioned ratchet pad
  • Largest ratcheting adjustment range
  • ANSI/ISEA Z89.1 type 1 class C compliant
  • CSA Z94.1 type 1 class C compliant
  • UL third-party tested and certified
  • Includes a marker clip for easy access to pens and markers
Catalog No. 50-006-87103 Supplier MSC Supplier No. 11641784
This hard hat features a 6-point ratcheting suspension and is designed to adapt to jobsite needs. It includes a BOLT™ Headlamp Mount, accessory slots, and a moisture-wicking sweatband for comfort.
  • Adjustable swinging ratchet for better comfort
  • Ratcheting suspension for quick adjustment
  • Six accessory slots for quick access
  • Moisture wicking sweatband to stay cool and dry fast
  • Secure accessories simultaneously
  • Cushioned ratchet pad
  • Largest ratcheting adjustment range
  • ANSI/ISEA Z89.1 type 1 class C compliant
  • CSA Z94.1 type 1 class C compliant
  • UL third-party tested and certified
  • Includes a marker clip for easy access to pens and markers