Learn More
MSC Milwaukee Inspection Camera, M12, 4 ft Cable, 360 Rotation, Kit
Catalog No. 5000534095
Encompass
The Milwaukee M12 M-Spector 360 4' Inspection Camera Kit is engineered to deliver clear visual inspections in challenging environments. It features a high-resolution camera and display, coupled with a flexible cable, making it ideal for a range of diagnostic tasks from drain lines to behind-wall inspections.
  • Clear images via 4.3" LCD screen and 720p camera
  • Flexible 4 ft cable for drain line and behind-wall inspections
  • 10 mm camera head allows access into tight spaces
  • 360 degree image rotation for flexible viewing angles
  • 3 levels of zoom to focus on specific points of interest
  • 5-mode brightness adjustment for optimized LED output
  • Share photos and videos using the included 32 GB SD card
  • Integrated into the M12 battery system for versatile power
  • Includes hook, mirror, magnet, and plumbing ball accessories
Catalog No. 50-005-34095 Supplier MSC Supplier No. 30598544
The Milwaukee M12 M-Spector 360 4' Inspection Camera Kit is engineered to deliver clear visual inspections in challenging environments. It features a high-resolution camera and display, coupled with a flexible cable, making it ideal for a range of diagnostic tasks from drain lines to behind-wall inspections.
  • Clear images via 4.3" LCD screen and 720p camera
  • Flexible 4 ft cable for drain line and behind-wall inspections
  • 10 mm camera head allows access into tight spaces
  • 360 degree image rotation for flexible viewing angles
  • 3 levels of zoom to focus on specific points of interest
  • 5-mode brightness adjustment for optimized LED output
  • Share photos and videos using the included 32 GB SD card
  • Integrated into the M12 battery system for versatile power
  • Includes hook, mirror, magnet, and plumbing ball accessories