Learn More
MSC Milwaukee M18 FUEL NEXUS Wet/Dry Vac, 6 gal., 1 ea
Catalog No. 5000803334
Encompass
The M18 FUEL NEXUS 6 gal. Wet/Dry Vacuum is designed for efficient cleanup with PACKOUT compatibility and VACLINK integration. This vacuum offers powerful suction and HEPA filtration for various applications.
  • Features a 6 gal. collection capacity
  • Provides a maximum air flow of 95 CFM
  • Equipped with HEPA filtration for fine dust collection
  • Bagless vacuum collection type
  • Durable plastic tank material
  • Includes a cord for power
  • Powered by battery for cordless operation
  • Delivers 3.5 hp for strong performance
Catalog No. 50-008-03334 Supplier MSC Supplier No. 56342553
The M18 FUEL NEXUS 6 gal. Wet/Dry Vacuum is designed for efficient cleanup with PACKOUT compatibility and VACLINK integration. This vacuum offers powerful suction and HEPA filtration for various applications.
  • Features a 6 gal. collection capacity
  • Provides a maximum air flow of 95 CFM
  • Equipped with HEPA filtration for fine dust collection
  • Bagless vacuum collection type
  • Durable plastic tank material
  • Includes a cord for power
  • Powered by battery for cordless operation
  • Delivers 3.5 hp for strong performance