Learn More
MSC Milwaukee Mesh Face Shield, Stainless Steel, 9.5 in., 1 ea
Catalog No. 5000684274
Encompass
This face shield mesh screen is engineered for use with Milwaukee Safety Helmets & Hard Hats, providing robust protection and a wide field of view for the user. It is constructed from highly durable stainless steel mesh and meets ANSI Z87.1+ standards.
  • Durable stainless steel mesh construction
  • Wide and clear field of view
  • Quick attachment and removal
  • Compatible with BOLT™ Front Slots
  • Meets ANSI Z87.1+ safety standards
  • Window dimensions: 9.5 in. high by 10 in. wide
Catalog No. 50-006-84274 Supplier MSC Supplier No. 82842378
This face shield mesh screen is engineered for use with Milwaukee Safety Helmets & Hard Hats, providing robust protection and a wide field of view for the user. It is constructed from highly durable stainless steel mesh and meets ANSI Z87.1+ standards.
  • Durable stainless steel mesh construction
  • Wide and clear field of view
  • Quick attachment and removal
  • Compatible with BOLT™ Front Slots
  • Meets ANSI Z87.1+ safety standards
  • Window dimensions: 9.5 in. high by 10 in. wide