Learn More
MSC Milwaukee Nitrile Gloves, Cut Level 1, SMARTSWIPE, Medium, 1 Pair
Catalog No. 5000792332
Encompass
These ANSI/ISEA 105-2016 Cut Level 1 Nitrile Dipped Gloves provide enhanced durability and protection. Featuring a soft, comfortable liner that wicks away moisture, they offer improved grip, dexterity, and mobility for handling small objects, making them ideal for material handling and general purpose applications.
  • Soft and comfortable liner
  • Sandy nitrile coating for better grip
  • Improved dexterity for easier mobility
  • Nylon/Lycra blend for moisture wicking
  • Touchscreen compatible fingertips
  • Breathable glove design
  • Color-coded band for easy cut level identification
  • ANSI/ISEA 105-2016 Cut Level 1 compliant
  • EN388 certified: Abrasion 3, Cut 1, Tearing 2, Puncture 1, TDM Cut A
  • Thirteen gauge knit
  • Elasticated knit cuff
Catalog No. 50-007-92332 Supplier MSC Supplier No. 77169415
These ANSI/ISEA 105-2016 Cut Level 1 Nitrile Dipped Gloves provide enhanced durability and protection. Featuring a soft, comfortable liner that wicks away moisture, they offer improved grip, dexterity, and mobility for handling small objects, making them ideal for material handling and general purpose applications.
  • Soft and comfortable liner
  • Sandy nitrile coating for better grip
  • Improved dexterity for easier mobility
  • Nylon/Lycra blend for moisture wicking
  • Touchscreen compatible fingertips
  • Breathable glove design
  • Color-coded band for easy cut level identification
  • ANSI/ISEA 105-2016 Cut Level 1 compliant
  • EN388 certified: Abrasion 3, Cut 1, Tearing 2, Puncture 1, TDM Cut A
  • Thirteen gauge knit
  • Elasticated knit cuff