Learn More
MSC Milwaukee Safety Vest, Class 2 Yellow Mesh, 4X/5X, 1 ea
Catalog No. 5000769431
Encompass
This high visibility safety vest provides a lightweight wear solution, featuring breathable mesh for comfort in hot working conditions. It includes one external chest pocket, one mic tab, and a secure zipper closure. This Class 2 high visibility reflective vest is recommended for environments with traffic traveling under 50 MPH or around heavy machinery, such as construction, road construction, traffic direction, utility crews, and survey crews.
  • Lightweight, breathable mesh
  • One chest pocket
  • Secure zipper closure
  • ANSI/ISEA 107-2020 compliant
  • Class 2 Type R
  • One mic tab
  • Dual sizing
Catalog No. 50-007-69431 Supplier MSC Supplier No. 06276265
This high visibility safety vest provides a lightweight wear solution, featuring breathable mesh for comfort in hot working conditions. It includes one external chest pocket, one mic tab, and a secure zipper closure. This Class 2 high visibility reflective vest is recommended for environments with traffic traveling under 50 MPH or around heavy machinery, such as construction, road construction, traffic direction, utility crews, and survey crews.
  • Lightweight, breathable mesh
  • One chest pocket
  • Secure zipper closure
  • ANSI/ISEA 107-2020 compliant
  • Class 2 Type R
  • One mic tab
  • Dual sizing