Learn More
MSC Milwaukee Tool Anchor Strap, 50 lb Capacity, D-Ring, 1 ea
Catalog No. 5000795881
Encompass
This anchor strap is designed to securely tether multiple tools while working at height. Featuring an oversized D-Ring that accommodates up to five carabiners, it is constructed with an abrasion-resistant 840D sleeve for enhanced durability. The strap's loop design allows it to securely wrap around beams and rails, while its green color-coding facilitates easy identification of its 50 lb weight rating.
  • Oversized D-ring holds 5 carabiners
  • Abrasion-resistant 840D sleeve
  • Loop securely wraps around beams and rails
  • Green color-coded for weight rating identification
  • Third-party tested at 2:1 safety factor
  • Maximum working capacity of 50 lb
  • ANSI/ISEA 121-2018 compliant
Catalog No. 50-007-95881 Supplier MSC Supplier No. 19942044
This anchor strap is designed to securely tether multiple tools while working at height. Featuring an oversized D-Ring that accommodates up to five carabiners, it is constructed with an abrasion-resistant 840D sleeve for enhanced durability. The strap's loop design allows it to securely wrap around beams and rails, while its green color-coding facilitates easy identification of its 50 lb weight rating.
  • Oversized D-ring holds 5 carabiners
  • Abrasion-resistant 840D sleeve
  • Loop securely wraps around beams and rails
  • Green color-coded for weight rating identification
  • Third-party tested at 2:1 safety factor
  • Maximum working capacity of 50 lb
  • ANSI/ISEA 121-2018 compliant