Learn More
MSC Milwaukee Tool AR/FR Vest, Class 2, Breakaway, Yellow, 1 ea
Catalog No. 5000792470
Encompass
This AR/FR CAT. 1 Class 2 Breakaway High Visibility Safety Vest is engineered for enhanced utility and protection. Featuring ten pockets, including pen pockets and a chest pocket with closure, it offers easy access to essential items. Constructed from lightweight WESTEX® DH material, it provides comfort and is rated to an ATPV of 5.7 cal./cm².
  • Quick external size adjustment
  • ATPV: 5.7 cal./cm²
  • Fall protection pass through
  • Secure hook and loop closure
  • Chest pocket with hook and loop closure
  • Five-point breakaway on shoulders, sides, and front
  • One internal pocket
  • Durable mic tabs
  • ANSI/ISEA 107-15 Type R Class 2 compliant
  • NFPA 2112 certified
  • ASTM F1506 compliant (Cat. 1)
  • Fabric weight: 6.5 oz/yd²
  • WESTEX® DH Fabric material
  • Ten total pockets
Catalog No. 50-007-92470 Supplier MSC Supplier No. 82854191
This AR/FR CAT. 1 Class 2 Breakaway High Visibility Safety Vest is engineered for enhanced utility and protection. Featuring ten pockets, including pen pockets and a chest pocket with closure, it offers easy access to essential items. Constructed from lightweight WESTEX® DH material, it provides comfort and is rated to an ATPV of 5.7 cal./cm².
  • Quick external size adjustment
  • ATPV: 5.7 cal./cm²
  • Fall protection pass through
  • Secure hook and loop closure
  • Chest pocket with hook and loop closure
  • Five-point breakaway on shoulders, sides, and front
  • One internal pocket
  • Durable mic tabs
  • ANSI/ISEA 107-15 Type R Class 2 compliant
  • NFPA 2112 certified
  • ASTM F1506 compliant (Cat. 1)
  • Fabric weight: 6.5 oz/yd²
  • WESTEX® DH Fabric material
  • Ten total pockets