Learn More
MSC Milwaukee Tool Battery Pack, M12 REDLITHIUM, 3.0 Ah, 12 Volt
Catalog No. NC3984154
Encompass
This compact battery pack provides superior power, runtime, and durability for cordless tools. Engineered with advanced electronics and construction, it offers extended life and efficient performance, even in extreme temperatures below 0°F/−18°C. Its compact design reduces user fatigue and allows access to tight work areas.
  • Delivers up to 3X more runtime and 20% more power compared to standard lithium-ion batteries.
  • REDLINK™ Intelligence ensures optimized performance and overload protection for the tool, battery, and charger.
  • All-weather performance provides fade-free power in extreme jobsite conditions.
  • Individual cell monitoring and temperature management maximize battery life.
  • Versatility: powers over 70 cordless solutions.
Catalog No. NC3984154 Supplier MSC Supplier No. 37812906
This compact battery pack provides superior power, runtime, and durability for cordless tools. Engineered with advanced electronics and construction, it offers extended life and efficient performance, even in extreme temperatures below 0°F/−18°C. Its compact design reduces user fatigue and allows access to tight work areas.
  • Delivers up to 3X more runtime and 20% more power compared to standard lithium-ion batteries.
  • REDLINK™ Intelligence ensures optimized performance and overload protection for the tool, battery, and charger.
  • All-weather performance provides fade-free power in extreme jobsite conditions.
  • Individual cell monitoring and temperature management maximize battery life.
  • Versatility: powers over 70 cordless solutions.