Learn More
MSC Milwaukee Tool BOLT Full Face Shield, Gasketed, Clear, 1 ea
Catalog No. 5000792041
Encompass
This BOLT gasketed full face shield provides superior overhead debris protection. It features a headlamp mount at the front, allowing seamless raising and lowering even with a headlamp attached. The lens has a fog-free interior coating for clear visibility and an anti-scratch exterior hard coating for durability in demanding job site conditions.
  • Best overhead debris protection
  • Gasket helps protect against falling debris
  • Raise and lower with headlamp mounted
  • Lasting fog-free interior coating
  • Anti-scratch hard exterior coating
  • Secure locking detents
  • Secures into front slots
  • Provides 180° field of view
  • Reversible adaptors allow use with both safety helmets and hard hats
  • Quick attachment and removal
  • Part of the BOLT accessory system for simultaneous accessory attachment
Catalog No. 50-007-92041 Supplier MSC Supplier No. 78406048
This BOLT gasketed full face shield provides superior overhead debris protection. It features a headlamp mount at the front, allowing seamless raising and lowering even with a headlamp attached. The lens has a fog-free interior coating for clear visibility and an anti-scratch exterior hard coating for durability in demanding job site conditions.
  • Best overhead debris protection
  • Gasket helps protect against falling debris
  • Raise and lower with headlamp mounted
  • Lasting fog-free interior coating
  • Anti-scratch hard exterior coating
  • Secure locking detents
  • Secures into front slots
  • Provides 180° field of view
  • Reversible adaptors allow use with both safety helmets and hard hats
  • Quick attachment and removal
  • Part of the BOLT accessory system for simultaneous accessory attachment