Learn More
MSC Milwaukee Tool Box Level, 48 in, Magnetic, Aluminum, 3 Vials
Catalog No. 5000527137
Encompass
The REDSTICK Box Level delivers industry-leading performance with best-in-class vial readability, frame durability, and magnet strength. Its ALL-METAL BACKBONE ensures the strongest and most durable frame and long-life accuracy. SHARPSITE Vial Technology provides superior readability with a magnified bubble and high-visibility vial spirit. Amplified Rare Earth Magnets maximize holding power.
  • All-metal backbone provides the strongest, most durable frame and reinforced vial retention.
  • SHARPSITE Vial Technology offers best-in-class readability and impact protection.
  • Guaranteed lifetime accuracy to .029° (.0005"/in, 0.5mm/M).
  • Amplified rare earth magnets maximize holding power.
  • High density, removable end-caps protect the frame and provide access in tight spaces.
  • Precision milled measuring surface guarantees accuracy in all working positions.
Catalog No. 50-005-27137 Supplier MSC Supplier No. 39610209
The REDSTICK Box Level delivers industry-leading performance with best-in-class vial readability, frame durability, and magnet strength. Its ALL-METAL BACKBONE ensures the strongest and most durable frame and long-life accuracy. SHARPSITE Vial Technology provides superior readability with a magnified bubble and high-visibility vial spirit. Amplified Rare Earth Magnets maximize holding power.
  • All-metal backbone provides the strongest, most durable frame and reinforced vial retention.
  • SHARPSITE Vial Technology offers best-in-class readability and impact protection.
  • Guaranteed lifetime accuracy to .029° (.0005"/in, 0.5mm/M).
  • Amplified rare earth magnets maximize holding power.
  • High density, removable end-caps protect the frame and provide access in tight spaces.
  • Precision milled measuring surface guarantees accuracy in all working positions.